Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snrpa
Entrez Gene ID
324121
Gene Name
small nuclear ribonucleoprotein polypeptide A
Gene Aliases
fc19d01, wu:fc19d01, zgc:77810
Chromosome Location
Chr.18: 35403042 - 35407695 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199671.1 NP_955965.1 2 - 3 116 106
GenBank mRNA BC064308.1 - 2 - 3 116 106

View Full Product Details