Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tagln3b
Entrez Gene ID
797861
Gene Name
transgelin 3b
Gene Aliases
fc80d07, tagln3, wu:fc80d07, zgc:66052
Chromosome Location
Chr.24: 21917211 - 21923873 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199720.1 NP_956014.1 1 - 2 213 109
GenBank mRNA BC062382.1 - 1 - 2 213 109

View Full Product Details