Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
psat1
Entrez Gene ID
327512
Gene Name
phosphoserine aminotransferase 1
Gene Aliases
fi15b02, wu:fi15b02, zgc:55738, zgc:77622
Chromosome Location
Chr.5: 69486341 - 69499615 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199819.2 NP_956113.1 2 - 3 243 73
GenBank mRNA BC044467.1 - 2 - 3 247 73
EH432914.1 - 2 - 3 275 73

View Full Product Details