Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pex26
Entrez Gene ID
334644
Gene Name
peroxisomal biogenesis factor 26
Gene Aliases
fk41g06, wu:fk41g06, zgc:64014
Chromosome Location
Chr.4: 5835442 - 5840625 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199920.1 NP_956214.1 3 - 4 798 145
GenBank mRNA BC053199.1 - 3 - 4 798 145
BC164102.1 - 3 - 4 732 145

View Full Product Details