Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
serp1
Entrez Gene ID
336797
Gene Name
stress-associated endoplasmic reticulum protein 1
Gene Aliases
RAMP4, wu:fb05f06, zgc:56419, zgc:86736
Chromosome Location
Chr.18: 3572401 - 3576434 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200034.1 NP_956328.1 2 - 3 263 73
GenBank mRNA BC049018.1 - 2 - 3 263 73
BC071411.1 - 2 - 3 246 73

View Full Product Details