The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_200254.1 | NP_956548.1 | 1 - 2 | 270 | 90 |
GenBank mRNA | BC048040.1 | - | 1 - 2 | 270 | 90 |
BC164392.1 | - | 1 - 2 | 165 | 90 |