Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snapc1b
Entrez Gene ID
393224
Gene Name
small nuclear RNA activating complex, polypeptide 1b
Gene Aliases
zgc:55475
Chromosome Location
Chr.13: 37586491 - 37600588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200254.1 NP_956548.1 8 - 9 1072 67
GenBank mRNA BC048040.1 - 8 - 9 1072 67
BC164392.1 - 8 - 9 967 67

View Full Product Details