Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
znhit3
Entrez Gene ID
393243
Gene Name
zinc finger, HIT-type containing 3
Gene Aliases
zgc:56688
Chromosome Location
Chr.5: 3927965 - 3937715 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200273.1 NP_956567.1 1 - 2 148 101
XM_021476032.1 XP_021331707.1 1 - 2 172 101
GenBank mRNA BC049063.1 - 1 - 2 148 101

View Full Product Details