Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
macrod2
Entrez Gene ID
393521
Gene Name
mono-ADP ribosylhydrolase 2
Gene Aliases
zgc:65960
Chromosome Location
Chr.13: 33703730 - 34560375 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200549.1 NP_956843.1 19 - 20 1394 59
XM_017358720.2 XP_017214209.1 20 - 21 1556 59
GenBank mRNA BC056529.1 - 19 - 20 1394 59
BC152658.1 - 19 - 20 1475 59

View Full Product Details