Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smfn
Entrez Gene ID
393717
Gene Name
small fragment nuclease
Gene Aliases
zgc:73191
Chromosome Location
Chr.10: 39893447 - 39905253 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200744.2 NP_957038.2 2 - 3 408 75
GenBank mRNA BC059540.1 - 2 - 3 292 75
EH454655.1 - 2 - 3 315 75
EH580645.1 - 2 - 3 442 75

View Full Product Details