Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smfn
Entrez Gene ID
393717
Gene Name
small fragment nuclease
Gene Aliases
zgc:73191
Chromosome Location
Chr.10: 39893447 - 39905253 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200744.2 NP_957038.2 4 - 5 600 91
GenBank mRNA BC059540.1 - 4 - 5 484 91
EH454655.1 - 4 - 5 507 91
EH463590.1 - 3 - 4 649 91
EH469615.1 - 3 - 4 634 91
EH580645.1 - 4 - 5 634 91

View Full Product Details