Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
chad
Entrez Gene ID
394038
Gene Name
chondroadherin
Gene Aliases
zgc:63475
Chromosome Location
Chr.12: 26512569 - 26525699 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201063.1 NP_957357.1 3 - 4 1172 136
GenBank mRNA BC054898.1 - 3 - 4 1172 136

View Full Product Details