Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc18a3b
Entrez Gene ID
394082
Gene Name
solute carrier family 18 member 3b
Gene Aliases
Slc18a3, VAChT-B, zgc:64220
Chromosome Location
Chr.12: 48958822 - 48960461 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201107.1 NP_957401.1 1 - 1 1449 86
GenBank mRNA BC054646.1 - 1 - 1 1449 86

View Full Product Details