Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
surf4
Entrez Gene ID
394102
Gene Name
surfeit 4
Gene Aliases
zgc:65806, zgc:77007
Chromosome Location
Chr.5: 1180363 - 1203004 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201127.1 NP_957421.1 3 - 4 436 97
GenBank mRNA BC056295.1 - 3 - 4 436 97
BC066509.1 - 3 - 4 422 97

View Full Product Details