The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201506.1 | NP_958914.1 | 8 - 9 | 745 | 85 |
GenBank mRNA | AY391421.1 | - | 8 - 9 | 696 | 85 |
BC044432.1 | - | 8 - 9 | 745 | 85 | |
BC065616.1 | - | 8 - 9 | 720 | 85 |