Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pwp2h
Entrez Gene ID
266990
Gene Name
PWP2 small subunit processome component
Gene Aliases
cb471, zgc:56063
Chromosome Location
Chr.1: 28899449 - 28950366 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213047.1 NP_998212.1 8 - 9 1103 72
GenBank mRNA BC047817.1 - 8 - 9 1103 72

View Full Product Details