Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
akr7a3
Entrez Gene ID
436642
Gene Name
aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase)
Gene Aliases
fd56g11, wu:fd56g11, zgc:92502
Chromosome Location
Chr.11: 41838801 - 41846060 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002369.1 NP_001002369.1 5 - 6 794 63
GenBank mRNA BC076040.1 - 5 - 6 794 63

View Full Product Details