Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
uqcrq
Entrez Gene ID
436768
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit VII
Gene Aliases
mg:bb03b10, zgc:92848
Chromosome Location
Chr.14: 141710 - 145074 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002495.3 NP_001002495.2 2 - 3 274 125
GenBank mRNA BC076310.1 - 2 - 3 245 125
BC165521.1 - - 193 125
EH545017.1 - 2 - 3 280 125

View Full Product Details