Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
htra3a
Entrez Gene ID
445108
Gene Name
HtrA serine peptidase 3a
Gene Aliases
CH1073-501A1.1, zgc:91963
Chromosome Location
Chr.1: 40741198 - 40758657 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003502.1 NP_001003502.1 6 - 7 1231 87
GenBank mRNA BC078402.1 - 6 - 7 1231 87

View Full Product Details