Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
chrne
Entrez Gene ID
445315
Gene Name
cholinergic receptor, nicotinic, epsilon
Gene Aliases
im:7154697, zgc:101010
Chromosome Location
Chr.5: 173955 - 184555 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001327883.1 NP_001314812.1 3 - 4 351 97
GenBank mRNA BC079485.1 - 3 - 4 322 97
JN242071.1 - 3 - 4 250 97

View Full Product Details