Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc35d1a
Entrez Gene ID
445400
Gene Name
solute carrier family 35 member D1a
Gene Aliases
slc35d1
Chromosome Location
Chr.6: 34848478 - 34860574 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003877.1 NP_001003877.1 6 - 7 503 120
GenBank mRNA AY648822.1 - 6 - 7 503 120
BC092185.1 - 6 - 7 477 120

View Full Product Details