Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mtdha
Entrez Gene ID
368910
Gene Name
metadherin a
Gene Aliases
lyric, lyricl, si:zc215i13.6, wu:fc33g06
Chromosome Location
Chr.19: 32029 - 50877 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007135.2 NP_001007136.2 1 - 2 423 102
XM_005159603.3 XP_005159660.1 1 - 2 496 102
GenBank mRNA BC068430.1 - 1 - 2 423 102
BC107640.1 - 1 - 2 497 102

View Full Product Details