Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kcnip1b
Entrez Gene ID
494089
Gene Name
Kv channel interacting protein 1 b
Gene Aliases
zgc:101531
Chromosome Location
Chr.10: 291705 - 326732 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008632.2 NP_001008632.2 1 - 2 197 115
GenBank mRNA BC086698.1 - 1 - 2 197 115

View Full Product Details