Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp2x6
Entrez Gene ID
541440
Gene Name
cytochrome P450, family 2, subfamily X, polypeptide 6
Gene Aliases
zgc:110749
Chromosome Location
Chr.25: 17360846 - 17367578 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013565.2 NP_001013583.1 1 - 2 203 122
GenBank mRNA BC078308.1 - 1 - 2 208 122
BC090406.1 - 1 - 2 207 122

View Full Product Details