Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ablim1b
- Entrez Gene ID
-
541550
- Gene Name
- actin binding LIM protein 1b
- Gene Aliases
-
si:dkeyp-84e12.1, zgc:112954
- Chromosome Location
-
Chr.12: 4690 - 44097 on Build GRCz11
View Full Product Details