Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppm1e
Entrez Gene ID
553417
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1E
Gene Aliases
-
Chromosome Location
Chr.10: 32683089 - 32788832 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020518.1 NP_001018354.1 3 - 4 584 76
GenBank mRNA AB113302.1 - 3 - 4 584 76
BC162507.1 - 3 - 4 572 76

View Full Product Details