The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001020762.1 | NP_001018598.1 | 5 - 6 | 744 | 66 |
XM_005156020.4 | XP_005156077.1 | 5 - 6 | 759 | 66 | |
GenBank mRNA | BC095686.1 | - | 5 - 6 | 744 | 66 |