Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab24
Entrez Gene ID
574423
Gene Name
RAB24, member RAS oncogene family
Gene Aliases
im:7142760, im:7155843, zgc:112047
Chromosome Location
Chr.21: 38732976 - 38740009 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029970.2 NP_001025141.1 8 - 9 1020 122
GenBank mRNA BC098610.1 - 8 - 9 953 122
EH563661.1 - 1 - 2 239 122

View Full Product Details