Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
snrnp35
Entrez Gene ID
569999
Gene Name
small nuclear ribonucleoprotein 35 (U11/U12)
Gene Aliases
zgc:112337
Chromosome Location
Chr.10: 44373349 - 44378142 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030241.1 NP_001025412.1 - 259 109
GenBank mRNA BC098619.1 - - 259 109

View Full Product Details