Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdc42l2
Entrez Gene ID
556226
Gene Name
cell division cycle 42 like 2
Gene Aliases
zgc:136923
Chromosome Location
Chr.23: 16955672 - 16961280 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039923.1 NP_001035012.1 2 - 3 316 85
XM_021469812.1 XP_021325487.1 2 - 3 624 85
XM_021469813.1 XP_021325488.1 2 - 3 609 85
XM_021469814.1 XP_021325489.1 2 - 3 624 85
GenBank mRNA BC114307.1 - 2 - 3 316 85

View Full Product Details