The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044709.2 | NP_001038174.2 | 16 - 17 | 1747 | 67 |
GenBank mRNA | BC139636.1 | - | 16 - 17 | 1747 | 67 |
DQ415957.1 | - | 16 - 17 | 1402 | 67 |