Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npr1a
Entrez Gene ID
560653
Gene Name
natriuretic peptide receptor 1a
Gene Aliases
sGC, si:dkey-27c15.1
Chromosome Location
Chr.19: 26423073 - 26627313 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044937.2 NP_001038402.1 8 - 9 1619 91
XM_005170050.4 XP_005170107.1 9 - 10 2299 91

View Full Product Details