Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snapin
Entrez Gene ID
567959
Gene Name
SNAP associated protein
Gene Aliases
si:dkey-222b8.2
Chromosome Location
Chr.19: 23919096 - 23922495 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045148.1 NP_001038613.1 3 - 4 282 80
GenBank mRNA BC139575.1 - 3 - 4 310 80

View Full Product Details