Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zbtb37
Entrez Gene ID
568471
Gene Name
zinc finger and BTB domain containing 37
Gene Aliases
si:dkey-192m14.2
Chromosome Location
Chr.22: 16027464 - 16036478 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045160.1 NP_001038625.1 2 - 3 1041 66
XM_005161718.4 XP_005161775.1 3 - 4 1970 66
GenBank mRNA BC163388.1 - 2 - 3 1067 66
BC163402.1 - 2 - 3 1067 66

View Full Product Details