Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lmo7b
Entrez Gene ID
558333
Gene Name
LIM domain 7b
Gene Aliases
DKEY-180K21.1, lmo7, zgc:152922
Chromosome Location
Chr.1: 3469 - 63711 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC122101.1 - 7 - 8 779 106

View Full Product Details