Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ftr82
Entrez Gene ID
338307
Gene Name
finTRIM family, member 82
Gene Aliases
cb579, id:ibd1340, sb:cb579, si:dkey-252h13.6, wu:fb99c05, zgc:153276
Chromosome Location
Chr.5: 30707129 - 30715229 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075103.1 NP_001068571.1 2 - 3 808 58
GenBank mRNA BC096901.1 - 1 - 2 89 58
BC124299.1 - 2 - 3 808 58

View Full Product Details