Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klf11b
Entrez Gene ID
559076
Gene Name
Kruppel-like factor 11b
Gene Aliases
zgc:154048
Chromosome Location
Chr.17: 32365941 - 32370025 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077604.1 NP_001071072.1 3 - 4 1191 64
XM_005158730.4 XP_005158787.1 3 - 4 1416 64
GenBank mRNA BC125942.1 - 3 - 4 1191 64
BC164539.1 - 3 - 4 1116 64

View Full Product Details