Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nptx2a
Entrez Gene ID
553230
Gene Name
neuronal pentraxin 2a
Gene Aliases
nptx2
Chromosome Location
Chr.3: 61565217 - 61593999 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109768.1 NP_001103238.1 2 - 3 1156 84
XM_005164291.4 XP_005164348.1 2 - 3 1878 84
XM_005164292.4 XP_005164349.1 2 - 3 1874 84
GenBank mRNA BC076490.1 - 2 - 3 708 84
BC115228.1 - 2 - 3 1156 84
FJ232033.1 - 2 - 3 653 84
KM036426.1 - 3 - 4 1554 84

View Full Product Details