Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nr2f5
Entrez Gene ID
30427
Gene Name
nuclear receptor subfamily 2, group F, member 5
Gene Aliases
COUP, couptf, svp46
Chromosome Location
Chr.16: 46294337 - 46327301 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131186.1 NP_571261.1 2 - 3 1296 61
GenBank mRNA BC162963.1 - 2 - 3 719 61
BC162999.1 - 2 - 3 719 61
X70300.1 - - 1296 61

View Full Product Details