Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bmp4
Entrez Gene ID
30612
Gene Name
bone morphogenetic protein 4
Gene Aliases
bmp-4, zbmp-4, zgc:100779
Chromosome Location
Chr.17: 50564103 - 50586135 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131342.2 NP_571417.1 3 - 4 689 106
GenBank mRNA BC078423.1 - 3 - 4 689 106
D49972.1 - 3 - 4 750 106
KM820425.1 - 1 - 2 356 106
U82231.1 - 1 - 2 356 106
U90122.1 - 3 - 4 696 106

View Full Product Details