Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
f7
Entrez Gene ID
114423
Gene Name
coagulation factor VII
Gene Aliases
wu:fb59f04, zgc:109870
Chromosome Location
Chr.1: 119960 - 125246 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131819.2 NP_571894.2 4 - 5 376 77
GenBank mRNA AY040345.1 - 4 - 5 360 77
BC095023.1 - 4 - 5 376 77
BC165195.1 - 4 - 5 394 77

View Full Product Details