Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
f7
Entrez Gene ID
114423
Gene Name
coagulation factor VII
Gene Aliases
wu:fb59f04, zgc:109870
Chromosome Location
Chr.1: 119960 - 125246 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131819.2 NP_571894.2 6 - 7 649 96
GenBank mRNA AY040345.1 - 6 - 7 633 96
BC095023.1 - 6 - 7 649 96
BC165195.1 - 6 - 7 667 96

View Full Product Details