Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmpoa
Entrez Gene ID
352907
Gene Name
thymopoietin a
Gene Aliases
LAP2, etID53005.4, tmpo, wu:fc26f12, wu:fd21f02, wu:fi29h05
Chromosome Location
Chr.4: 16857722 - 16876343 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328563.1 NP_001315492.1 1 - 2 300 69
NM_001328564.1 NP_001315493.1 1 - 2 300 69
NM_001328565.1 NP_001315494.1 1 - 2 300 69
NM_180968.3 NP_851299.1 1 - 2 300 69
XM_005164720.4 XP_005164777.1 1 - 2 307 69
XM_005164721.4 XP_005164778.1 1 - 2 306 69
XM_005164722.4 XP_005164779.1 1 - 2 306 69
GenBank mRNA AJ320189.1 - 1 - 2 248 69
AJ320190.1 - 1 - 2 248 69
AJ320191.1 - 1 - 2 248 69
AJ320192.1 - 1 - 2 246 69
BC048056.1 - 1 - 2 300 69
BC165860.1 - 1 - 2 276 69

View Full Product Details