Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crabp2a
Entrez Gene ID
171480
Gene Name
cellular retinoic acid binding protein 2, a
Gene Aliases
cb432, cb434, crabp2, sb:cb434, wu:fc51a11, zgc:110496
Chromosome Location
Chr.16: 14383479 - 14396984 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182859.1 NP_878279.1 1 - 2 348 82
GenBank mRNA AF497478.1 - 1 - 2 348 82
BC091960.1 - 1 - 2 369 82
BC164791.1 - 1 - 2 119 82

View Full Product Details