Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
add3a
Entrez Gene ID
556762
Gene Name
adducin 3 (gamma) a
Gene Aliases
add3, wu:fb38g03, wu:fc16e06, zgc:55560, zgc:55992, zgc:77226
Chromosome Location
Chr.22: 30047220 - 30272394 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199663.1 NP_955957.1 14 - 15 1818 71
XM_005161804.4 XP_005161861.1 11 - 12 1824 71
XM_005161805.4 XP_005161862.1 11 - 12 1824 71
XM_005161807.4 XP_005161864.1 11 - 12 1823 71
XM_017353146.2 XP_017208635.1 11 - 12 1825 71
XM_017353147.2 XP_017208636.1 12 - 13 1695 71
XM_017353148.2 XP_017208637.1 11 - 12 2043 71
XM_017353149.2 XP_017208638.1 11 - 12 1825 71
XM_017353150.2 XP_017208639.1 11 - 12 1824 71
XM_021469329.1 XP_021325004.1 13 - 14 1792 71
XM_021469330.1 XP_021325005.1 11 - 12 1553 71
XM_021469331.1 XP_021325006.1 13 - 14 1791 71
GenBank mRNA AY391406.1 - 10 - 11 1532 71
BC045384.1 - 11 - 12 1722 71
BC050486.1 - 14 - 15 1818 71
BC165643.1 - 10 - 11 1539 71

View Full Product Details