Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lrrc42
Entrez Gene ID
326793
Gene Name
leucine rich repeat containing 42
Gene Aliases
wu:fe17g08, zgc:56446, zgc:77847
Chromosome Location
Chr.2: 26604773 - 26611033 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199747.2 NP_956041.2 5 - 6 1083 81
GenBank mRNA BC049443.1 - 5 - 6 962 81
BC062861.1 - 5 - 6 1038 81

View Full Product Details