Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc1a2b
Entrez Gene ID
335836
Gene Name
solute carrier family 1 member 2b
Gene Aliases
eaat2, fj34b12, slc1a2, wu:fj34b12, zgc:65897
Chromosome Location
Chr.25: 37366698 - 37407757 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199979.2 NP_956273.2 2 - 3 393 67
XM_009298220.3 XP_009296495.1 2 - 3 178 67
XM_009298221.3 XP_009296496.1 2 - 3 157 67
XM_009298222.3 XP_009296497.1 2 - 3 344 67
GenBank mRNA BC056751.1 - 2 - 3 318 67
BC155822.1 - 2 - 3 344 67
HM138691.1 - 1 - 2 135 67

View Full Product Details