Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ucp3
Entrez Gene ID
794081
Gene Name
uncoupling protein 3
Gene Aliases
ucp2l, zgc:63967
Chromosome Location
Chr.10: 36650235 - 36656696 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC053173.1 - 7 - 8 1079 77

View Full Product Details