Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arhgap4a
Entrez Gene ID
393416
Gene Name
Rho GTPase activating protein 4a
Gene Aliases
zgc:63672
Chromosome Location
Chr.23: 24989387 - 25015614 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200444.1 NP_956738.1 12 - 13 1403 106
XM_005162155.4 XP_005162212.1 12 - 13 1503 106
GenBank mRNA BC054931.1 - 12 - 13 1403 106

View Full Product Details