Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pabpc1b
Entrez Gene ID
393856
Gene Name
poly A binding protein, cytoplasmic 1 b
Gene Aliases
pabpc1, zgc:77608
Chromosome Location
Chr.19: 12121944 - 12145764 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200882.1 NP_957176.1 9 - 10 1794 56
XM_021468020.1 XP_021323695.1 8 - 9 1711 56
GenBank mRNA BC059662.1 - 9 - 10 1660 56
BC063948.1 - 9 - 10 1794 56

View Full Product Details